Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BBS1
Entrez Gene ID
582
Gene Name
Bardet-Biedl syndrome 1
Gene Aliases
BBS2L2
Chromosome Location
Chr.11: 66510648 - 66533613 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024649.4 NP_078925.3 10 - 11 968 71
GenBank mRNA AF503941.1 - 10 - 11 968 71
AK095638.1 - 9 - 10 975 71
AK296311.1 - 6 - 7 683 71
BC047642.1 - 9 - 10 927 71
BC109064.2 - 10 - 11 961 71
BX647612.1 - 10 - 11 997 71

View Full Product Details