The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134771.1 | NP_001128243.1 | 10 - 11 | 1478 | 74 |
NM_020708.4 | NP_065759.1 | 10 - 11 | 1496 | 74 | |
XM_017027981.1 | XP_016883470.1 | 10 - 11 | 1478 | 74 | |
GenBank mRNA | AB033002.1 | - | 10 - 11 | 1311 | 74 |
AF208159.1 | - | 10 - 11 | 1333 | 74 | |
AK289758.1 | - | 10 - 11 | 1455 | 74 | |
AK294059.1 | - | 6 - 7 | 673 | 74 | |
AK295900.1 | - | 4 - 5 | 692 | 74 | |
BC132668.1 | - | 10 - 11 | 1421 | 74 | |
BC132670.1 | - | 10 - 11 | 1421 | 74 |