Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DZANK1
Entrez Gene ID
55184
Gene Name
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
Chromosome Location
Chr.20: 18383367 - 18467185 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005260743.3 XP_005260800.1 5 - 6 646 72
XM_006723584.2 XP_006723647.1 5 - 6 685 72
XM_011529275.2 XP_011527577.1 5 - 6 1396 72
XM_017027916.1 XP_016883405.1 5 - 6 1396 72
XM_017027917.1 XP_016883406.1 7 - 8 249
XM_017027918.1 XP_016883407.1 7 - 8 249
XM_017027919.1 XP_016883408.1 7 - 8 249
XM_017027920.1 XP_016883409.1 5 - 6 782 72
GenBank mRNA AK055544.1 - 5 - 6 535 72
BC036231.1 - 5 - 6 731 72
CR749327.1 - 7 - 8 249

View Full Product Details