The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258330.1 | NP_001245259.1 | 13 - 14 | 1532 | 73 |
NM_001258331.1 | NP_001245260.1 | 12 - 13 | 1432 | 73 | |
NM_177996.2 | NP_818932.1 | 12 - 13 | 1427 | 73 | |
XM_011528681.1 | XP_011526983.1 | 11 - 12 | 1459 | 73 | |
XM_011528682.1 | XP_011526984.1 | 11 - 12 | 1456 | 73 | |
XM_017027713.1 | XP_016883202.1 | 12 - 13 | 1489 | 73 | |
XM_017027714.1 | XP_016883203.1 | 12 - 13 | 1486 | 73 | |
XM_017027715.1 | XP_016883204.1 | 11 - 12 | 1456 | 73 | |
XM_017027719.1 | XP_016883208.1 | 12 - 13 | 1461 | 73 | |
XM_017027720.1 | XP_016883209.1 | 14 - 15 | 1720 | 73 | |
GenBank mRNA | AK096848.1 | - | 12 - 13 | 1479 | 73 |
AK126875.1 | - | 13 - 14 | 1530 | 73 | |
BC013885.2 | - | 12 - 13 | 1432 | 73 |