The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023037.2 | NP_075463.2 | 36 - 37 | 5262 | 86 |
XM_006719749.3 | XP_006719812.1 | 36 - 37 | 6343 | 86 | |
XM_017020303.1 | XP_016875792.1 | 36 - 37 | 6820 | 86 | |
XM_017020304.1 | XP_016875793.1 | 36 - 37 | 5262 | 86 | |
XM_017020305.1 | XP_016875794.1 | 36 - 37 | 6839 | 86 | |
XM_017020306.1 | XP_016875795.1 | 15 - 16 | 1981 | 86 | |
XM_017020307.1 | XP_016875796.1 | 9 - 10 | 1334 | 86 | |
GenBank mRNA | AL049784.1 | - | 36 - 37 | 5259 | 86 |
U50534.1 | - | 36 - 37 | 5261 | 86 |