Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRY
Entrez Gene ID
10129
Gene Name
FRY microtubule binding protein
Gene Aliases
13CDNA73, 214K23.2, C13orf14, CG003, bA207N4.2, bA37E23.1
Chromosome Location
Chr.13: 32029742 - 32299122 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023037.2 NP_075463.2 47 - 48 7318 120
XM_006719749.3 XP_006719812.1 47 - 48 8399 120
XM_017020303.1 XP_016875792.1 47 - 48 8876 120
XM_017020304.1 XP_016875793.1 47 - 48 7318 120
XM_017020305.1 XP_016875794.1 47 - 48 8895 120
XM_017020306.1 XP_016875795.1 26 - 27 4037 120
XM_017020307.1 XP_016875796.1 20 - 21 3390 120
GenBank mRNA AK091422.1 - 2 - 3 156 120
AL049784.1 - 47 - 48 7315 120

View Full Product Details