Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRY
Entrez Gene ID
10129
Gene Name
FRY microtubule binding protein
Gene Aliases
13CDNA73, 214K23.2, C13orf14, CG003, bA207N4.2, bA37E23.1
Chromosome Location
Chr.13: 32029742 - 32299122 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023037.2 NP_075463.2 57 - 58 8882 147
XM_006719749.3 XP_006719812.1 58 - 59 9972 147
XM_017020303.1 XP_016875792.1 58 - 59 10449 147
XM_017020304.1 XP_016875793.1 58 - 59 8891 147
XM_017020305.1 XP_016875794.1 58 - 59 10468 147
XM_017020306.1 XP_016875795.1 37 - 38 5610 147
XM_017020307.1 XP_016875796.1 31 - 32 4963 147
GenBank mRNA AK091422.1 - 13 - 14 1729 147
AL049784.1 - 57 - 58 8879 147
U50534.1 - 58 - 59 8890 147
U57961.1 - 2 - 3 244 148

View Full Product Details