The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023037.2 | NP_075463.2 | 31 - 32 | 4517 | 102 |
XM_006719749.3 | XP_006719812.1 | 31 - 32 | 5598 | 102 | |
XM_017020303.1 | XP_016875792.1 | 31 - 32 | 6075 | 102 | |
XM_017020304.1 | XP_016875793.1 | 31 - 32 | 4517 | 102 | |
XM_017020305.1 | XP_016875794.1 | 31 - 32 | 6094 | 102 | |
XM_017020306.1 | XP_016875795.1 | 10 - 11 | 1236 | 102 | |
XM_017020307.1 | XP_016875796.1 | 4 - 5 | 589 | 102 |