The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004685.3 | NP_004676.3 | 10 - 11 | 1464 | 138 |
XM_011535306.1 | XP_011533608.1 | 11 - 12 | 1524 | 138 | |
XM_011535307.1 | XP_011533609.1 | 11 - 12 | 1524 | 138 | |
XM_017020845.1 | XP_016876334.1 | 10 - 11 | 1410 | 138 | |
XM_017020846.1 | XP_016876335.1 | 7 - 8 | 729 | 138 | |
GenBank mRNA | AF072928.1 | - | 6 - 7 | 678 | 138 |
AF406619.1 | - | 10 - 11 | 1145 | 138 | |
AK314587.1 | - | 10 - 11 | 1329 | 138 | |
AL832017.2 | - | 10 - 11 | 1387 | 138 | |
BC040012.1 | - | 10 - 11 | 1336 | 138 | |
BU177677.1 | - | 5 - 6 | 529 | 138 |