The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004685.3 | NP_004676.3 | 8 - 9 | 1288 | 136 |
XM_011535306.1 | XP_011533608.1 | 9 - 10 | 1348 | 136 | |
XM_011535307.1 | XP_011533609.1 | 9 - 10 | 1348 | 136 | |
XM_017020845.1 | XP_016876334.1 | 8 - 9 | 1234 | 136 | |
XM_017020846.1 | XP_016876335.1 | 5 - 6 | 553 | 136 | |
GenBank mRNA | AK314587.1 | - | 8 - 9 | 1153 | 136 |
AL832017.2 | - | 8 - 9 | 1211 | 136 | |
BC040012.1 | - | 8 - 9 | 1160 | 136 |