The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015078.3 | NP_055893.3 | 20 - 21 | 2593 | 101 |
XM_011512585.2 | XP_011510887.1 | 12 - 13 | 2030 | 101 | |
XM_011512586.2 | XP_011510888.1 | 13 - 14 | 1444 | 101 | |
XM_017005943.1 | XP_016861432.1 | 20 - 21 | 3314 | 101 | |
GenBank mRNA | AB020668.1 | - | 17 - 18 | 1905 | 101 |
AY172737.1 | - | 19 - 20 | 2145 | 101 | |
BX649003.1 | - | - | 6772 | 101 |