The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130081.2 | NP_001123553.1 | 14 - 15 | 1670 | 123 |
NM_002662.4 | NP_002653.1 | 14 - 15 | 1670 | 123 | |
XM_005247533.2 | XP_005247590.1 | 14 - 15 | 1739 | 123 | |
XM_005247534.2 | XP_005247591.1 | 14 - 15 | 1739 | 123 | |
XM_011512897.1 | XP_011511199.1 | 14 - 15 | 1739 | 123 | |
XM_011512898.1 | XP_011511200.1 | 14 - 15 | 1739 | 123 | |
XM_011512899.2 | XP_011511201.1 | 4 - 5 | 822 | 123 | |
XM_017006623.1 | XP_016862112.1 | 14 - 15 | 1670 | 123 | |
XM_017006624.1 | XP_016862113.1 | 3 - 4 | 944 | 123 | |
GenBank mRNA | AB209907.1 | - | 14 - 15 | 1614 | 123 |
BC068976.1 | - | 14 - 15 | 1621 | 123 | |
BX648900.1 | - | 1 - 2 | 199 | 123 | |
U38545.1 | - | 14 - 15 | 1638 | 123 |