The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130081.2 | NP_001123553.1 | 1 - 2 | 99 | 99 |
NM_002662.4 | NP_002653.1 | 1 - 2 | 99 | 99 | |
XM_017006623.1 | XP_016862112.1 | 1 - 2 | 99 | 99 | |
GenBank mRNA | DA325526.1 | - | 1 - 2 | 88 | 99 |
DA464847.1 | - | 1 - 2 | 99 | 99 |