The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130081.2 | NP_001123553.1 | 21 - 22 | 2559 | 79 |
NM_002662.4 | NP_002653.1 | 22 - 23 | 2673 | 79 | |
XM_005247533.2 | XP_005247590.1 | 22 - 23 | 2742 | 79 | |
XM_005247534.2 | XP_005247591.1 | 21 - 22 | 2628 | 79 | |
XM_011512897.1 | XP_011511199.1 | 22 - 23 | 2742 | 79 | |
XM_011512898.1 | XP_011511200.1 | 22 - 23 | 2742 | 79 | |
XM_011512899.2 | XP_011511201.1 | 12 - 13 | 1825 | 79 | |
XM_017006623.1 | XP_016862112.1 | 21 - 22 | 2559 | 79 | |
XM_017006624.1 | XP_016862113.1 | 11 - 12 | 1947 | 79 | |
GenBank mRNA | AB209907.1 | - | 21 - 22 | 2503 | 79 |
AJ276230.1 | - | 4 - 5 | 334 | 79 | |
BC068976.1 | - | 22 - 23 | 2624 | 79 | |
BX648900.1 | - | 8 - 9 | 1088 | 79 | |
U38545.1 | - | 22 - 23 | 2641 | 79 |