The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130081.2 | NP_001123553.1 | 2 - 3 | 293 | 129 |
NM_002662.4 | NP_002653.1 | 2 - 3 | 293 | 129 | |
XM_005247533.2 | XP_005247590.1 | 2 - 3 | 362 | 129 | |
XM_005247534.2 | XP_005247591.1 | 2 - 3 | 362 | 129 | |
XM_011512897.1 | XP_011511199.1 | 2 - 3 | 362 | 129 | |
XM_011512898.1 | XP_011511200.1 | 2 - 3 | 362 | 129 | |
XM_017006623.1 | XP_016862112.1 | 2 - 3 | 293 | 129 | |
GenBank mRNA | AB209907.1 | - | 2 - 3 | 237 | 129 |
BC068976.1 | - | 2 - 3 | 244 | 129 | |
DA325526.1 | - | 2 - 3 | 282 | 129 | |
DA464847.1 | - | 2 - 3 | 293 | 129 | |
U38545.1 | - | 2 - 3 | 261 | 129 |