The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276342.1 | NP_001263271.1 | 5 - 6 | 664 | 70 |
NM_001318707.1 | NP_001305636.1 | 4 - 5 | 579 | 70 | |
NM_001318708.1 | NP_001305637.1 | 6 - 7 | 723 | 70 | |
NM_001318709.1 | NP_001305638.1 | 5 - 6 | 664 | 70 | |
NM_013326.4 | NP_037458.3 | 7 - 8 | 808 | 70 | |
XM_011525939.1 | XP_011524241.1 | 6 - 7 | 680 | 70 | |
XM_017025717.1 | XP_016881206.1 | 5 - 6 | 617 | 70 | |
GenBank mRNA | AK057192.1 | - | 7 - 8 | 769 | 70 |
AK294169.1 | - | 4 - 5 | 526 | 70 | |
AK295246.1 | - | 6 - 7 | 680 | 70 | |
AK296230.1 | - | 5 - 6 | 615 | 70 | |
BC002950.1 | - | 7 - 8 | 733 | 70 | |
BC008305.1 | - | 7 - 8 | 740 | 70 |