The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004164.2 | NP_004155.2 | 3 - 4 | 101 | |
GenBank mRNA | AI076320.1 | - | 1 - 2 | 297 | 101 |
AK291978.1 | - | 3 - 4 | 101 | ||
AW772327.1 | - | 1 - 2 | 285 | 101 |