Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABI3BP
Entrez Gene ID
25890
Gene Name
ABI family member 3 binding protein
Gene Aliases
NESHBP, TARSH
Chromosome Location
Chr.3: 100749335 - 100993508 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015429.3 NP_056244.2 22 - 23 1972 75
XM_005247281.2 XP_005247338.1 54 - 55 4054 75
XM_005247282.2 XP_005247339.1 53 - 54 4012 75
XM_005247283.2 XP_005247340.1 53 - 54 4006 75
XM_005247284.2 XP_005247341.1 53 - 54 3997 75
XM_005247285.2 XP_005247342.1 53 - 54 3994 75
XM_005247287.2 XP_005247344.1 53 - 54 3988 75
XM_005247288.2 XP_005247345.1 53 - 54 3982 75
XM_005247289.2 XP_005247346.1 53 - 54 3979 75
XM_005247290.2 XP_005247347.1 54 - 55 4054 75
XM_005247293.2 XP_005247350.1 52 - 53 3946 75
XM_005247294.2 XP_005247351.1 52 - 53 3937 75
XM_005247295.2 XP_005247352.1 52 - 53 3916 75
XM_005247296.2 XP_005247353.1 52 - 53 3913 75
XM_005247298.2 XP_005247355.1 52 - 53 3910 75
XM_005247299.3 XP_005247356.1 52 - 53 3922 75
XM_005247300.2 XP_005247357.1 51 - 52 3880 75
XM_005247302.2 XP_005247359.1 51 - 52 3850 75
XM_005247303.2 XP_005247360.1 50 - 51 3793 75
XM_005247305.2 XP_005247362.1 49 - 50 3709 75
XM_005247306.2 XP_005247363.1 47 - 48 3646 75
XM_005247309.1 XP_005247366.1 52 - 53 3687 75
XM_006713570.2 XP_006713633.1 51 - 52 3868 75
XM_006713571.2 XP_006713634.1 51 - 52 3838 75
XM_006713572.2 XP_006713635.1 51 - 52 3832 75
XM_011512639.1 XP_011510941.1 53 - 54 3994 75
XM_011512640.1 XP_011510942.1 53 - 54 3988 75
XM_011512641.1 XP_011510943.1 53 - 54 3979 75
XM_011512643.1 XP_011510945.1 52 - 53 3973 75
XM_011512644.1 XP_011510946.1 53 - 54 3970 75
XM_011512645.1 XP_011510947.1 53 - 54 3970 75
XM_011512646.1 XP_011510948.1 53 - 54 3961 75
XM_011512647.1 XP_011510949.1 52 - 53 3919 75
XM_011512648.1 XP_011510950.1 52 - 53 3916 75
XM_011512649.1 XP_011510951.1 52 - 53 3910 75
XM_011512651.1 XP_011510953.1 51 - 52 3853 75
XM_011512652.1 XP_011510954.1 51 - 52 3841 75
XM_011512653.1 XP_011510955.1 50 - 51 3832 75
XM_011512654.1 XP_011510956.1 51 - 52 3823 75
XM_011512656.1 XP_011510958.1 50 - 51 3757 75
XM_011512657.1 XP_011510959.1 51 - 52 3829 75
XM_011512658.1 XP_011510960.1 49 - 50 3742 75
XM_011512659.1 XP_011510961.1 49 - 50 3721 75
XM_011512660.1 XP_011510962.1 49 - 50 3691 75
XM_017006105.1 XP_016861594.1 53 - 54 3997 75
XM_017006106.1 XP_016861595.1 53 - 54 3979 75
XM_017006107.1 XP_016861596.1 52 - 53 3922 75
XM_017006108.1 XP_016861597.1 49 - 50 3757 75
GenBank mRNA AB056106.1 - 4 - 5 408 75
AK093861.1 - 4 - 5 231 75
AK123737.1 - 14 - 15 999 75
AK123748.1 - 12 - 13 788 75
AK299938.1 - 32 - 33 2054 75
BC030221.1 - 22 - 23 1952 75
BX648726.1 - 30 - 31 2061 75

View Full Product Details