Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABI3BP
Entrez Gene ID
25890
Gene Name
ABI family member 3 binding protein
Gene Aliases
NESHBP, TARSH
Chromosome Location
Chr.3: 100749335 - 100993508 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015429.3 NP_056244.2 23 - 24 2052 70
XM_005247281.2 XP_005247338.1 55 - 56 4134 70
XM_005247282.2 XP_005247339.1 54 - 55 4092 70
XM_005247283.2 XP_005247340.1 54 - 55 4086 70
XM_005247284.2 XP_005247341.1 54 - 55 4077 70
XM_005247285.2 XP_005247342.1 54 - 55 4074 70
XM_005247287.2 XP_005247344.1 54 - 55 4068 70
XM_005247288.2 XP_005247345.1 54 - 55 4062 70
XM_005247289.2 XP_005247346.1 54 - 55 4059 70
XM_005247290.2 XP_005247347.1 55 - 56 4134 70
XM_005247293.2 XP_005247350.1 53 - 54 4026 70
XM_005247294.2 XP_005247351.1 53 - 54 4017 70
XM_005247295.2 XP_005247352.1 53 - 54 3996 70
XM_005247296.2 XP_005247353.1 53 - 54 3993 70
XM_005247298.2 XP_005247355.1 53 - 54 3990 70
XM_005247299.3 XP_005247356.1 53 - 54 4002 70
XM_005247300.2 XP_005247357.1 52 - 53 3960 70
XM_005247302.2 XP_005247359.1 52 - 53 3930 70
XM_005247303.2 XP_005247360.1 51 - 52 3873 70
XM_005247305.2 XP_005247362.1 50 - 51 3789 70
XM_005247306.2 XP_005247363.1 48 - 49 3726 70
XM_005247309.1 XP_005247366.1 53 - 54 3767 70
XM_006713570.2 XP_006713633.1 52 - 53 3948 70
XM_006713571.2 XP_006713634.1 52 - 53 3918 70
XM_006713572.2 XP_006713635.1 52 - 53 3912 70
XM_011512639.1 XP_011510941.1 54 - 55 4074 70
XM_011512640.1 XP_011510942.1 54 - 55 4068 70
XM_011512641.1 XP_011510943.1 54 - 55 4059 70
XM_011512643.1 XP_011510945.1 53 - 54 4053 70
XM_011512644.1 XP_011510946.1 54 - 55 4050 70
XM_011512645.1 XP_011510947.1 54 - 55 4050 70
XM_011512646.1 XP_011510948.1 54 - 55 4041 70
XM_011512647.1 XP_011510949.1 53 - 54 3999 70
XM_011512648.1 XP_011510950.1 53 - 54 3996 70
XM_011512649.1 XP_011510951.1 53 - 54 3990 70
XM_011512651.1 XP_011510953.1 52 - 53 3933 70
XM_011512652.1 XP_011510954.1 52 - 53 3921 70
XM_011512653.1 XP_011510955.1 51 - 52 3912 70
XM_011512654.1 XP_011510956.1 52 - 53 3903 70
XM_011512656.1 XP_011510958.1 51 - 52 3837 70
XM_011512657.1 XP_011510959.1 52 - 53 3909 70
XM_011512658.1 XP_011510960.1 50 - 51 3822 70
XM_011512659.1 XP_011510961.1 50 - 51 3801 70
XM_011512660.1 XP_011510962.1 50 - 51 3771 70
XM_017006105.1 XP_016861594.1 54 - 55 4077 70
XM_017006106.1 XP_016861595.1 54 - 55 4059 70
XM_017006107.1 XP_016861596.1 53 - 54 4002 70
XM_017006108.1 XP_016861597.1 50 - 51 3837 70
GenBank mRNA AB056106.1 - 5 - 6 488 70
AK093861.1 - 5 - 6 311 70
AK123737.1 - 15 - 16 1079 70
AK123748.1 - 13 - 14 868 70
AK299938.1 - 33 - 34 2134 70
BC030221.1 - 23 - 24 2032 70
BX648726.1 - 31 - 32 2141 70

View Full Product Details