Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABI3BP
Entrez Gene ID
25890
Gene Name
ABI family member 3 binding protein
Gene Aliases
NESHBP, TARSH
Chromosome Location
Chr.3: 100749335 - 100993508 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015429.3 NP_056244.2 1 - 2 180 72
XM_005247281.2 XP_005247338.1 1 - 2 183 72
XM_005247282.2 XP_005247339.1 1 - 2 183 72
XM_005247283.2 XP_005247340.1 1 - 2 183 72
XM_005247284.2 XP_005247341.1 1 - 2 183 72
XM_005247285.2 XP_005247342.1 1 - 2 183 72
XM_005247287.2 XP_005247344.1 1 - 2 183 72
XM_005247288.2 XP_005247345.1 1 - 2 183 72
XM_005247289.2 XP_005247346.1 1 - 2 183 72
XM_005247290.2 XP_005247347.1 1 - 2 183 72
XM_005247291.2 XP_005247348.1 1 - 2 183 72
XM_005247293.2 XP_005247350.1 1 - 2 183 72
XM_005247294.2 XP_005247351.1 1 - 2 183 72
XM_005247295.2 XP_005247352.1 1 - 2 183 72
XM_005247296.2 XP_005247353.1 1 - 2 183 72
XM_005247298.2 XP_005247355.1 1 - 2 183 72
XM_005247299.3 XP_005247356.1 1 - 2 198 72
XM_005247300.2 XP_005247357.1 1 - 2 183 72
XM_005247301.2 XP_005247358.1 1 - 2 183 72
XM_005247302.2 XP_005247359.1 1 - 2 183 72
XM_005247303.2 XP_005247360.1 1 - 2 183 72
XM_005247305.2 XP_005247362.1 1 - 2 183 72
XM_005247306.2 XP_005247363.1 1 - 2 183 72
XM_006713569.2 XP_006713632.1 1 - 2 183 72
XM_006713570.2 XP_006713633.1 1 - 2 183 72
XM_006713571.2 XP_006713634.1 1 - 2 183 72
XM_006713572.2 XP_006713635.1 1 - 2 183 72
XM_011512639.1 XP_011510941.1 1 - 2 183 72
XM_011512640.1 XP_011510942.1 1 - 2 183 72
XM_011512641.1 XP_011510943.1 1 - 2 183 72
XM_011512643.1 XP_011510945.1 1 - 2 183 72
XM_011512644.1 XP_011510946.1 1 - 2 183 72
XM_011512645.1 XP_011510947.1 1 - 2 183 72
XM_011512646.1 XP_011510948.1 1 - 2 183 72
XM_011512647.1 XP_011510949.1 1 - 2 183 72
XM_011512648.1 XP_011510950.1 1 - 2 183 72
XM_011512649.1 XP_011510951.1 1 - 2 183 72
XM_011512651.1 XP_011510953.1 1 - 2 183 72
XM_011512652.1 XP_011510954.1 1 - 2 183 72
XM_011512653.1 XP_011510955.1 1 - 2 183 72
XM_011512654.1 XP_011510956.1 1 - 2 183 72
XM_011512656.1 XP_011510958.1 1 - 2 183 72
XM_011512657.1 XP_011510959.1 1 - 2 183 72
XM_011512658.1 XP_011510960.1 1 - 2 183 72
XM_011512659.1 XP_011510961.1 1 - 2 183 72
XM_011512660.1 XP_011510962.1 1 - 2 183 72
XM_017006105.1 XP_016861594.1 1 - 2 183 72
XM_017006106.1 XP_016861595.1 1 - 2 183 72
XM_017006107.1 XP_016861596.1 1 - 2 183 72
XM_017006108.1 XP_016861597.1 1 - 2 183 72
GenBank mRNA AK025204.1 - 1 - 2 140 72
AL833204.1 - 1 - 2 143 72
BC030221.1 - 1 - 2 160 72
DA342832.1 - 1 - 2 180 72

View Full Product Details