Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ABI3BP
- Entrez Gene ID
-
25890
- Gene Name
- ABI family member 3 binding protein
- Gene Aliases
-
NESHBP, TARSH
- Chromosome Location
-
Chr.3: 100749335 - 100993508 on Build GRCh38
View Full Product Details