The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193534.1 | NP_001180463.1 | 1 - 2 | 206 | 82 |
NM_012161.3 | NP_036293.1 | 1 - 2 | 206 | 82 | |
NR_036464.1 | 1 - 2 | 206 | 82 | ||
GenBank mRNA | AF142481.1 | - | 1 - 2 | 128 | 82 |
AF157323.1 | - | 1 - 2 | 67 | 82 | |
AF176700.1 | - | 1 - 2 | 90 | 82 | |
AF199420.1 | - | 1 - 2 | 95 | 82 | |
AK310860.1 | - | 1 - 2 | 135 | 82 | |
BC012320.1 | - | 1 - 2 | 135 | 82 | |
BC030656.1 | - | 1 - 2 | 177 | 82 | |
BI464829.1 | - | 1 - 2 | 185 | 82 | |
DB080171.1 | - | 1 - 2 | 207 | 82 |