The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193535.1 | NP_001180464.1 | 4 - 5 | 656 | 97 |
NM_012161.3 | NP_036293.1 | 4 - 5 | 707 | 97 | |
NR_036464.1 | 5 - 6 | 811 | 97 | ||
XM_006713959.3 | XP_006714022.1 | 5 - 6 | 803 | 97 | |
XM_011513831.2 | XP_011512133.1 | 4 - 5 | 489 | 97 | |
XM_011513832.2 | XP_011512134.1 | 4 - 5 | 636 | 97 | |
XM_011513833.2 | XP_011512135.1 | 3 - 4 | 447 | 97 | |
XM_017008019.1 | XP_016863508.1 | 6 - 7 | 1574 | 97 | |
GenBank mRNA | AF142481.1 | - | 4 - 5 | 629 | 97 |
AF157323.1 | - | 4 - 5 | 568 | 97 | |
AF174591.1 | - | 1 - 2 | 114 | 97 | |
AF176700.1 | - | 4 - 5 | 591 | 97 | |
AK000153.1 | - | 3 - 4 | 417 | 97 | |
AK295510.1 | - | 4 - 5 | 626 | 97 | |
AK310860.1 | - | 4 - 5 | 636 | 97 | |
AL832808.1 | - | 2 - 3 | 343 | 97 | |
BC012320.1 | - | 5 - 6 | 740 | 97 | |
BC030656.1 | - | 4 - 5 | 678 | 97 | |
BX537957.1 | - | 4 - 5 | 602 | 97 |