The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018227.5 | NP_060697.4 | 24 - 25 | 2242 | 82 |
XM_017008359.1 | XP_016863848.1 | 24 - 25 | 2219 | 82 | |
GenBank mRNA | AB014773.1 | - | 24 - 25 | 2190 | 82 |
AK001670.1 | - | 6 - 7 | 591 | 82 | |
AK094164.1 | - | 16 - 17 | 1448 | 82 | |
AK314371.1 | - | 24 - 25 | 2219 | 82 | |
AL832015.2 | - | 24 - 25 | 2211 | 82 | |
AL832458.2 | - | 24 - 25 | 2211 | 82 | |
AY359880.1 | - | 11 - 12 | 1563 | 82 | |
BC126484.1 | - | 24 - 25 | 2237 | 82 | |
BC126486.1 | - | 24 - 25 | 2237 | 82 | |
EF623993.1 | - | 24 - 25 | 2190 | 82 |