The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160147.1 | NP_001153619.1 | 12 - 13 | 2689 | 77 |
NM_030637.2 | NP_085140.2 | 11 - 12 | 2668 | 77 | |
XM_011537189.2 | XP_011535491.1 | 13 - 14 | 2780 | 77 | |
XM_017021668.1 | XP_016877157.1 | 12 - 13 | 2759 | 77 | |
XM_017021669.1 | XP_016877158.1 | 10 - 11 | 2528 | 77 | |
GenBank mRNA | AB051492.1 | - | 8 - 9 | 3029 | 77 |
AK058137.1 | - | 9 - 10 | 1371 | 77 | |
AK125372.1 | - | 12 - 13 | 2571 | 77 | |
BC030703.2 | - | 11 - 12 | 2622 | 77 | |
DQ315474.1 | - | 12 - 13 | 2181 | 77 |