The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006544.3 | NP_006535.1 | 3 - 4 | 518 | 104 |
XM_005267272.3 | XP_005267329.1 | 3 - 4 | 603 | 104 | |
GenBank mRNA | AK295085.1 | - | 3 - 4 | 425 | 104 |
AK312523.1 | - | 3 - 4 | 523 | 104 | |
BC041126.1 | - | 3 - 4 | 484 | 104 | |
BI460915.1 | - | 3 - 4 | 529 | 104 | |
U85946.1 | - | 3 - 4 | 462 | 104 |