The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006544.3 | NP_006535.1 | 8 - 9 | 964 | 96 |
XM_005267272.3 | XP_005267329.1 | 8 - 9 | 1049 | 96 | |
GenBank mRNA | AK295085.1 | - | 8 - 9 | 871 | 96 |
AK303531.1 | - | 7 - 8 | 707 | 96 | |
AK312523.1 | - | 8 - 9 | 969 | 96 | |
BC041126.1 | - | 8 - 9 | 930 | 96 | |
U85946.1 | - | 8 - 9 | 908 | 96 |