The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100164.1 | NP_001093634.1 | 7 - 8 | 1638 | 72 |
NM_001100165.1 | NP_001093635.1 | 6 - 7 | 1398 | 72 | |
NM_001100166.1 | NP_001093636.1 | 6 - 7 | 1296 | 72 | |
NM_014721.2 | NP_055536.2 | 7 - 8 | 1536 | 72 | |
GenBank mRNA | AB014580.1 | - | 7 - 8 | 1453 | 72 |
AK297799.1 | - | 7 - 8 | 1443 | 72 | |
AK298263.1 | - | 7 - 8 | 1638 | 72 | |
AK300221.1 | - | 6 - 7 | 1291 | 72 | |
BC150630.1 | - | 7 - 8 | 1443 | 72 | |
BC152416.1 | - | 7 - 8 | 1453 | 72 | |
CR749345.1 | - | 6 - 7 | 1336 | 72 |