The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005743.1 | NP_001005743.1 | 6 - 7 | 549 | 113 |
NM_001005744.1 | NP_001005744.1 | 6 - 7 | 549 | 113 | |
NM_001320114.1 | NP_001307043.1 | 6 - 7 | 569 | 113 | |
GenBank mRNA | AF015040.1 | - | 6 - 7 | 507 | 113 |
AF108092.1 | - | 4 - 5 | 334 | 113 | |
AF171938.1 | - | 6 - 7 | 499 | 113 | |
AF171939.1 | - | 6 - 7 | 499 | 113 | |
AK304193.1 | - | 4 - 5 | 352 | 113 | |
BX248073.1 | - | 5 - 6 | 406 | 113 | |
BX647137.1 | - | 6 - 7 | 508 | 113 | |
EU265734.1 | - | 4 - 5 | 268 | 113 | |
EU265736.1 | - | 4 - 5 | 268 | 113 |