Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GJA9
- Entrez Gene ID
-
81025
- Gene Name
- gap junction protein alpha 9
- Gene Aliases
-
CX58, CX59, GJA10
- Chromosome Location
-
Chr.1: 38874067 - 38881626 on Build GRCh38
- Gene Symbol
- GJA9-MYCBP
- Entrez Gene ID
-
100527950
- Gene Name
- GJA9-MYCBP readthrough
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 38862490 - 38881626 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037633.1 |
|
1 - 2 |
199 |
141 |
NR_037634.1 |
|
1 - 2 |
199 |
141 |
GenBank mRNA |
AK123051.1 |
- |
1 - 2 |
193 |
141 |
BC051675.1 |
- |
1 - 2 |
170 |
141 |
View Full Product Details