The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031275.4 | NP_112565.1 | 1 - 2 | 109 | 116 |
GenBank mRNA | BC029506.1 | - | 1 - 2 | 109 | 116 |
BI561514.1 | - | 1 - 2 | 113 | 116 |