Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAPN12
Entrez Gene ID
147968
Gene Name
calpain 12
Gene Aliases
-
Chromosome Location
Chr.19: 38730124 - 38744474 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144691.4 NP_653292.2 9 - 10 1437 76
XM_017026355.1 XP_016881844.1 9 - 10 1437 76
XM_017026356.1 XP_016881845.1 9 - 10 1437 76
XM_017026357.1 XP_016881846.1 9 - 10 1437 76
XM_017026358.1 XP_016881847.1 9 - 10 1437 76
XM_017026359.1 XP_016881848.1 9 - 10 1437 76
XM_017026360.1 XP_016881849.1 9 - 10 1437 76
XM_017026361.1 XP_016881850.1 9 - 10 1437 76
XM_017026362.1 XP_016881851.1 9 - 10 1437 76
XM_017026363.1 XP_016881852.1 9 - 10 1437 76
XM_017026364.1 XP_016881853.1 9 - 10 1073 76
XM_017026365.1 XP_016881854.1 9 - 10 1437 76
GenBank mRNA AK127398.1 - 9 - 10 1437 76

View Full Product Details