Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDH10
Entrez Gene ID
1008
Gene Name
cadherin 10
Gene Aliases
-
Chromosome Location
Chr.5: 24487100 - 24644978 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317222.1 NP_001304151.1 3 - 4 1032 95
NM_001317224.1 NP_001304153.1 2 - 3 533 95
NM_006727.4 NP_006718.2 3 - 4 1032 95
XM_011513923.2 XP_011512225.1 4 - 5 1071 95
XM_017008915.1 XP_016864404.1 3 - 4 990 95
GenBank mRNA AB035303.1 - 3 - 4 856 95
BC144168.1 - 3 - 4 834 95
DN993415.1 - 2 - 3 545 95
KJ534806.1 - 2 - 3 524 95
KJ534808.1 - 2 - 3 524 95

View Full Product Details