The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317222.1 | NP_001304151.1 | 4 - 5 | 1160 | 94 |
NM_001317224.1 | NP_001304153.1 | 3 - 4 | 661 | 94 | |
NM_006727.4 | NP_006718.2 | 4 - 5 | 1160 | 94 | |
XM_011513923.2 | XP_011512225.1 | 5 - 6 | 1199 | 94 | |
XM_017008915.1 | XP_016864404.1 | 4 - 5 | 1118 | 94 | |
GenBank mRNA | AB035303.1 | - | 4 - 5 | 984 | 94 |
BC144168.1 | - | 4 - 5 | 962 | 94 | |
KJ534806.1 | - | 3 - 4 | 652 | 94 | |
KJ534808.1 | - | 3 - 4 | 652 | 94 |