The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317222.1 | NP_001304151.1 | 5 - 6 | 1324 | 98 |
NM_001317224.1 | NP_001304153.1 | 4 - 5 | 825 | 98 | |
NM_006727.4 | NP_006718.2 | 5 - 6 | 1324 | 98 | |
XM_011513923.2 | XP_011512225.1 | 6 - 7 | 1363 | 98 | |
XM_017008915.1 | XP_016864404.1 | 5 - 6 | 1282 | 98 | |
GenBank mRNA | AB035303.1 | - | 5 - 6 | 1148 | 98 |
BC144168.1 | - | 5 - 6 | 1126 | 98 | |
KJ534806.1 | - | 4 - 5 | 816 | 98 | |
KJ534808.1 | - | 4 - 5 | 816 | 98 |