The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317222.1 | NP_001304151.1 | 9 - 10 | 1992 | 113 |
NM_001317224.1 | NP_001304153.1 | 9 - 10 | 1630 | 113 | |
NM_006727.4 | NP_006718.2 | 10 - 11 | 2129 | 113 | |
XM_011513923.2 | XP_011512225.1 | 11 - 12 | 2168 | 113 | |
XM_017008915.1 | XP_016864404.1 | 10 - 11 | 2087 | 113 | |
GenBank mRNA | AB035303.1 | - | 10 - 11 | 1953 | 113 |
BC144168.1 | - | 9 - 10 | 1794 | 113 | |
KJ534806.1 | - | 9 - 10 | 1621 | 113 | |
KJ534808.1 | - | 9 - 10 | 1621 | 113 |