Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NGF
Entrez Gene ID
4803
Gene Name
nerve growth factor
Gene Aliases
Beta-NGF, HSAN5, NGFB
Chromosome Location
Chr.1: 115285915 - 115338253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002506.2 NP_002497.2 1 - 2 32 86
GenBank mRNA BC032517.2 - 1 - 2 37 86
X52599.1 - 1 - 2 32 86

View Full Product Details