Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CEP170
- Entrez Gene ID
-
9859
- Gene Name
- centrosomal protein 170
- Gene Aliases
-
FAM68A, KAB, KIAA0470
- Chromosome Location
-
Chr.1: 243124428 - 243255936 on Build GRCh38
View Full Product Details