The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007351.2 | NP_031377.2 | 4 - 5 | 1024 | 70 |
XM_017007893.1 | XP_016863382.1 | 5 - 6 | 1265 | 70 | |
GenBank mRNA | AK125557.1 | - | 5 - 6 | 971 | 70 |
AK302421.1 | - | 4 - 5 | 403 | 70 | |
AK313566.1 | - | 4 - 5 | 1049 | 70 | |
BC063848.1 | - | 4 - 5 | 1024 | 70 | |
U27109.1 | - | 4 - 5 | 1024 | 70 |