The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306146.1 | NP_001293075.1 | 5 - 6 | 920 | 115 |
NM_002112.3 | NP_002103.2 | 5 - 6 | 920 | 115 | |
XM_017022094.1 | XP_016877583.1 | 5 - 6 | 684 | 115 | |
XM_017022095.1 | XP_016877584.1 | 5 - 6 | 684 | 115 | |
XM_017022096.1 | XP_016877585.1 | 4 - 5 | 469 | 115 | |
XM_017022097.1 | XP_016877586.1 | 3 - 4 | 767 | 115 | |
XM_017022098.1 | XP_016877587.1 | 2 - 3 | 287 | 115 | |
XM_017022099.1 | XP_016877588.1 | 5 - 6 | 684 | 115 | |
GenBank mRNA | AK308833.1 | - | 5 - 6 | 920 | 115 |
AK310479.1 | - | 3 - 4 | 446 | 115 | |
AK311072.1 | - | 5 - 6 | 639 | 115 | |
BC130527.1 | - | 5 - 6 | 621 | 115 | |
BC144173.1 | - | 5 - 6 | 621 | 115 | |
M60445.1 | - | 5 - 6 | 629 | 115 | |
X54297.1 | - | 5 - 6 | 680 | 115 |