Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDC
Entrez Gene ID
3067
Gene Name
histidine decarboxylase
Gene Aliases
-
Chromosome Location
Chr.15: 50241945 - 50266049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306146.1 NP_001293075.1 8 - 9 1296 62
NM_002112.3 NP_002103.2 8 - 9 1296 62
XM_017022094.1 XP_016877583.1 8 - 9 1165 62
XM_017022095.1 XP_016877584.1 8 - 9 1165 62
XM_017022096.1 XP_016877585.1 7 - 8 950 62
XM_017022097.1 XP_016877586.1 6 - 7 1248 62
XM_017022098.1 XP_016877587.1 5 - 6 768 62
XM_017022099.1 XP_016877588.1 8 - 9 1165 62
GenBank mRNA AK027221.1 - 1 - 2 699 62
BC130527.1 - 8 - 9 997 62
BC144173.1 - 8 - 9 997 62
M60445.1 - 8 - 9 1005 62
X54297.1 - 8 - 9 1056 62

View Full Product Details