The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005805.5 | NP_005796.1 | 10 - 11 | 1235 | 129 |
GenBank mRNA | AB209220.1 | - | 3 - 4 | 4411 | 129 |
AK055128.1 | - | 10 - 11 | 1472 | 129 | |
BC009524.1 | - | 1 - 2 | 125 | 129 | |
BC066336.1 | - | 10 - 11 | 1081 | 129 | |
BM978668.1 | - | 2 - 3 | 129 | ||
U86782.1 | - | 10 - 11 | 967 | 129 |