The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138376.2 | NP_612385.2 | 5 - 6 | 701 | 72 |
GenBank mRNA | AK074553.1 | - | 5 - 6 | 684 | 72 |
BC008647.1 | - | 5 - 6 | 642 | 72 | |
BC030822.1 | - | 5 - 6 | 645 | 72 | |
BC053538.1 | - | 5 - 6 | 649 | 72 | |
BX248066.1 | - | 5 - 6 | 473 | 72 | |
BX640732.1 | - | 5 - 6 | 580 | 72 | |
BX648584.1 | - | 5 - 6 | 656 | 72 |