Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC5
Entrez Gene ID
91875
Gene Name
tetratricopeptide repeat domain 5
Gene Aliases
Strap
Chromosome Location
Chr.14: 20289131 - 20305994 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138376.2 NP_612385.2 8 - 9 1111 89
GenBank mRNA AK074553.1 - 8 - 9 1094 89
BC008647.1 - 8 - 9 1052 89
BC030822.1 - 8 - 9 1055 89
BC053538.1 - 8 - 9 1059 89
BX248066.1 - 8 - 9 883 89
BX640732.1 - 8 - 9 990 89

View Full Product Details