The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182502.3 | NP_872308.2 | 7 - 8 | 820 | 156 |
XM_011531608.2 | XP_011529910.1 | 7 - 8 | 820 | 156 | |
GenBank mRNA | AK290904.1 | - | 7 - 8 | 820 | 156 |
AL833167.1 | - | 7 - 8 | 818 | 156 | |
BC126195.1 | - | 7 - 8 | 742 | 156 | |
BX537945.1 | - | 7 - 8 | 818 | 156 |