The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182502.3 | NP_872308.2 | 8 - 9 | 1083 | 139 |
XM_011531608.2 | XP_011529910.1 | 8 - 9 | 1083 | 139 | |
GenBank mRNA | AK290904.1 | - | 8 - 9 | 1083 | 139 |
AL833167.1 | - | 8 - 9 | 1081 | 139 | |
BC126195.1 | - | 8 - 9 | 1005 | 139 | |
BX537945.1 | - | 8 - 9 | 1081 | 139 |