The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256588.1 | NP_001243517.1 | 1 - 2 | 104 | |
NM_014579.3 | NP_055394.2 | 1 - 2 | 104 | ||
GenBank mRNA | AF186081.1 | - | 1 - 2 | 255 | 104 |
AK314974.1 | - | 1 - 2 | 104 | ||
BC096723.1 | - | 1 - 2 | 142 | 104 | |
BC098254.1 | - | 1 - 2 | 142 | 104 | |
BC110057.1 | - | 1 - 2 | 142 | 104 | |
BP359281.1 | - | 1 - 2 | 104 | ||
CN306082.1 | - | 1 - 2 | 237 | 104 | |
D83039.1 | - | 1 - 2 | 249 | 104 | |
DA871745.1 | - | 1 - 2 | 104 |