The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031485.3 | NP_113673.3 | 4 - 5 | 913 | 82 |
GenBank mRNA | AF337808.1 | - | 4 - 5 | 692 | 82 |
AK074676.1 | - | 4 - 5 | 881 | 82 | |
AK300225.1 | - | 3 - 4 | 531 | 82 | |
AK315625.1 | - | 4 - 5 | 705 | 82 | |
AL713729.1 | - | 4 - 5 | 688 | 82 | |
BC002440.2 | - | 4 - 5 | 688 | 82 | |
BC036281.1 | - | 4 - 5 | 664 | 82 | |
BC047351.1 | - | 4 - 5 | 664 | 82 | |
EF011614.1 | - | 4 - 5 | 686 | 82 |