Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASZ1
Entrez Gene ID
136991
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
Chromosome Location
Chr.7: 117363222 - 117427523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301821.1 NP_001288750.1 12 - 13 1312 151
NM_001301822.1 NP_001288751.1 11 - 12 1227 151
NM_130768.2 NP_570124.1 12 - 13 1339 151
GenBank mRNA AF461259.1 - 12 - 13 1305 151
AK093445.1 - 12 - 13 1292 151
BC028695.1 - 11 - 12 2455 151
BC034963.1 - 11 - 12 2461 151
BC068488.1 - 10 - 11 2346 151
BC126186.1 - 12 - 13 1298 151
BC126188.1 - 12 - 13 1298 151
BC144212.1 - 12 - 13 1271 151

View Full Product Details