Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBXN8
Entrez Gene ID
7993
Gene Name
UBX domain protein 8
Gene Aliases
D8S2298E, REP8, UBXD6
Chromosome Location
Chr.8: 30729131 - 30767006 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282189.1 NP_001269118.1 4 - 5 321 93
NM_005671.3 NP_005662.1 4 - 5 432 93
XM_011544655.2 XP_011542957.1 6 - 7 1509 93
XM_017013862.1 XP_016869351.1 5 - 6 621 93
GenBank mRNA AK296600.1 - 4 - 5 606 93
AY302140.1 - 4 - 5 296 93
BC020694.1 - 4 - 5 416 93
BI462115.1 - 4 - 5 437 93
D83767.1 - 4 - 5 422 93

View Full Product Details